Learn More
Sino Biological Recombinant Human HSF1 Protein (His Tag)
Supplier:  Sino Biological 12245H07E100
Encompass_Preferred
A DNA sequence encoding the human HSF1 isoform long (Q00613-1) (Asp 2-Ser 529) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7917