Learn More
Sino Biological Recombinant Human CLEC2B Protein (hFc Tag)
Supplier:  Sino Biological 12255H01H100
Encompass_Preferred
A DNA sequence encoding the human CLEC2B (AAH05254.1) (Lys26-His149) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-7923