Learn More
Sino Biological Human KRAS / K-Ras (Q61H) Protein (His Tag)
Supplier:  Sino Biological 12259H07E50
Encompass_Preferred
A DNA sequence encoding the human KRAS (P01116-2) (Thr2-Cys185, with natural variant Gln 61 His ) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7930