Learn More
Sino Biological Recombinant Human IL-24 Protein (mFc Tag)
Supplier:  Sino Biological 12265H05H50
Encompass_Preferred
A DNA sequence encoding the human IL24 (NP_001172085.1) (Met1-Leu207) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-7934