Learn More
Sino Biological Recombinant Human IL-24 Protein (His Tag)
Supplier:  Sino Biological 12265H08H50
Encompass_Preferred
A DNA sequence encoding the human IL24 isoform 3 (NP_001172085.1) (Met 1-Leu 207) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7936