Learn More
Sino Biological Recombinant Human CD47 Protein (ECD, hFc Tag), HPLC-verified
Catalog No. 501617943
Encompass_Preferred
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7943 Supplier Sino Biological Supplier No. 12283H02H100
Add to Cart
Add to Cart
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with the Fc region of human IgG1 at the C-terminus.