Learn More
Cayman Chemical 123 TrIdocosaheaenyl Glyc 50mg
SDP
Catalog No. 502964269
Encompass_Preferred
A monoclonal antibody-derived tetrapeptide that acts as a partial agonist at the glycine site of the NMDA receptor active in vivo and readily passes the BBB blocks neuropathic pain enhances learning and memory and significantly reduces anxiety in animals
Catalog No. 50-296-4269 Supplier Cayman Chemical Supplier No. 2315250 MG
A monoclonal antibody-derived tetrapeptide that acts as a partial agonist at the glycine site of the NMDA receptor active in vivo and readily passes the BBB blocks neuropathic pain enhances learning and memory and significantly reduces anxiety in animals