Learn More
Cayman Chemical 123 TrIlnolaIdoyl GlycRo 50mg
SDP
Catalog No. 502941714
Encompass_Preferred
A triacylglycerol; dietary administration lowers serum levels of TXB2, PGF2, and PGE in rats from 0.6-6.3% w/w
Catalog No. 50-294-1714 Supplier Cayman Chemical Supplier No. 2694850 MG
A triacylglycerol; dietary administration lowers serum levels of TXB2, PGF2, and PGE in rats from 0.6-6.3% w/w