Learn More
Sino Biological Recombinant Human MICA Protein (ECD, His Tag), MICA*018, HPLC-verified
Supplier:  Sino Biological 12302H08H100
Encompass_Preferred
A DNA sequence encoding the human MICA (AAH16929.1) extracellular domain (Met 1-Gln 308) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7956