Learn More
Sino Biological Recombinant Human Netrin G1/NTNG1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12313H08H50
Encompass_Preferred
A DNA sequence encoding the human NTNG1 (NP_055732.2) (Met1-Gly408) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7961