Learn More
Sino Biological Recombinant Human IL-29/IFN-lambda 1 Protein (His Tag)
Supplier:  Sino Biological 12339H08H100
Encompass_Preferred
A DNA sequence encoding the human IL29 (NP_742152.1) (Met 1-Thr 200) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7962