Learn More
AdipoGen 1233B (1 mg)
SDP
Supplier:  AdipoGen AGCN20112M001
Encompass_Preferred
Chemical. CAS: 34668-61-6. Formula: C18H30O6. MW: 342.4. Isolated from Scopulariopsis sp. Hydroxy-acid of antibiotic 1233A. Antibiotic. Antibacterial.
Catalog No. 50-205-3475