Learn More
Cayman Chemical 1,2,3,4,6,7,8,9-Octachlorodibenzofuran, Purity: ≥95%, CAS: 39001-02-0, 10 mg
SDP
Catalog No. 502918974
Encompass_Preferred
A dioxin-like polychlorinated dibenzofuran; induces a seven-fold increase in EROD activity in mouse liver microsomes but not in skin or lung homogenates; selectively accumulates and persists in the liver over adipose tissue in monkeys; has been found in the soil near municipal waste incinerators, in freshwater fish, and in blood from individuals that lived near or in herbicide-sprayed regions
Catalog No. 50-291-8974 Supplier Cayman Chemical Supplier No. 3862410 MG
A dioxin-like polychlorinated dibenzofuran; induces a seven-fold increase in EROD activity in mouse liver microsomes but not in skin or lung homogenates; selectively accumulates and persists in the liver over adipose tissue in monkeys; has been found in the soil near municipal waste incinerators, in freshwater fish, and in blood from individuals that lived near or in herbicide-sprayed regions