Learn More
Sigma Aldrich Fine Chemicals Biosciences 1234-Tetrahydronaphthalene1L
Catalog No. 502898059
Encompass
1 2 3 4-Tetrahydronaphthalene (Tetralin(TM)) a bicyclic hydrocarbon is a constituent of petroleum and coal tar. It is used in coal liquefaction and as an alternative to turpentine in paints and waxes. Degradation of tetralin by Corynebacterium sp. strain C125 and by pyrolysis have been reported. A study reports the transformation of naphthalene to tetralin in the presence of activated carbon supported molybdenum carbides. Dehydrogenation of tetralin by microwave heating in the presence of platinum-supported activated carbon has been investigated.
Catalog No. 50-289-8059 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5226511L
Add to Cart
Add to Cart
1 2 3 4-Tetrahydronaphthalene (Tetralin(TM)) a bicyclic hydrocarbon is a constituent of petroleum and coal tar. It is used in coal liquefaction and as an alternative to turpentine in paints and waxes. Degradation of tetralin by Corynebacterium sp. strain C125 and by pyrolysis have been reported. A study reports the transformation of naphthalene to tetralin in the presence of activated carbon supported molybdenum carbides. Dehydrogenation of tetralin by microwave heating in the presence of platinum-supported activated carbon has been investigated.