Learn More
Sino Biological Recombinant Human IL-28A/Interferon lambda-2 Protein (His Tag)
Supplier:  Sino Biological 12340H08Y100
Encompass_Preferred
A DNA sequence encoding the human IFNL2 (NP_742150.1) (Val26-Val200) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7967