Learn More
Sino Biological Recombinant Human FGF-17 Protein
Supplier:  Sino Biological 12342HNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of human FGF17 (O60258-1) (Thr23-Thr216) was expressed, with a N-terminal Met.
Catalog No. 50-161-7973