Learn More
Sino Biological Recombinant Human KLK15 Protein (hFc Tag)
Supplier:  Sino Biological 12343H02H100
Encompass_Preferred
A DNA sequence encoding the human KLK15 (NP_059979.2) (Met1-Asn256) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7975