Learn More
Sino Biological Recombinant Human LYPLA2 Protein (His Tag)
Supplier:  Sino Biological 12360H07E100
Encompass_Preferred
A DNA sequence encoding the human LYPLA2 (O95372) (Met 1-Val 231) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7979