Learn More
Cayman Chemical 123678 HexaCLOrodIbenzo 2.5mg
SDP
Catalog No. 502918752
Encompass_Preferred
A dioxin-like polychlorinated dibenzofuran; induces expression of the genes encoding AHH and EROD in H-4-II-E rat hepatoma cells (EC50s = 1.47 and 1.24 nM, respectively); reduces weight, induces thymic atrophy, and induces the expression of genes encoding CYP1A1 and 4-chlorobiphenyl hydroxylase in rats (ED50s = 3.2, 0.9, 0.35, and 0.21 µmol/kg, respectively)
Catalog No. 50-291-8752 Supplier Cayman Chemical Supplier No. 386212.5 MG
A dioxin-like polychlorinated dibenzofuran; induces expression of the genes encoding AHH and EROD in H-4-II-E rat hepatoma cells (EC50s = 1.47 and 1.24 nM, respectively); reduces weight, induces thymic atrophy, and induces the expression of genes encoding CYP1A1 and 4-chlorobiphenyl hydroxylase in rats (ED50s = 3.2, 0.9, 0.35, and 0.21 µmol/kg, respectively)