Learn More
Sino Biological Recombinant Human Casein Kinase 1 gamma 2 Protein (His Tag)
Supplier:  Sino Biological 12373H07B50
Encompass_Preferred
A DNA sequence encoding the mature form of human CSNK1G2 (P78368-1) (Met 18-Lys 415) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7984