Learn More
Sino Biological Recombinant Human AK3L1 Protein (His & GST Tag)
Supplier:  Sino Biological 12406H20B50
Encompass_Preferred
A DNA sequence encoding the human AK4 (P27144-1) (Ala 2-Tyr 223) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7989