Learn More
Sino Biological Recombinant Human CD58/LFA-3 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 12409H02H100
Encompass_Preferred
A DNA sequence encoding the human CD58 (Q9BRW0) (Met1-Arg215) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7993