Learn More
Sino Biological Recombinant Human C1D Protein (GST Tag)
Supplier:  Sino Biological 12432H09E100
Encompass_Preferred
A DNA sequence encoding the human C1D (Q13901) (Met 1-Ser 141) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8003