Learn More
Sino Biological Recombinant Human UBE2T Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12440H08E50
Encompass_Preferred
A DNA sequence encoding the human UBE2T (Q9NPD8) (Met1-Val197) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8010