Learn More
Sino Biological Recombinant Human RhoA Protein (His Tag)
Supplier:  Sino Biological 12441H07B100
Encompass_Preferred
A DNA sequence encoding the human RHOA (P61586-1) (Met 1-Leu 193) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8011