Learn More
Sino Biological Recombinant Human Adenylate Kinase 1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12442H07E50
Encompass_Preferred
A DNA sequence encoding the human AK1 (AAH01116.1) (Met 1-Lys 194) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8013