Learn More
Sino Biological Recombinant Human DUSP14 Protein (His & MBP Tag)
Supplier:  Sino Biological 12443H10E100
Encompass_Preferred
A DNA sequence encoding the human DUSP14 (O95147) (Met 1 –His 191) was fused with an N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-161-8014