Learn More
Sino Biological Recombinant Human CD300A/LMIR1 Protein (hFc Tag)
Supplier:  Sino Biological 12449H02H100
Encompass_Preferred
A DNA sequence encoding the human CD300A (Q9UGN4-1)(Met1-Gln178) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8016