Learn More
Sino Biological Recombinant Human CD300A/LMIR1 Protein (His Tag)
Supplier:  Sino Biological 12449H08H100
Encompass_Preferred
A DNA sequence encoding the human CD300A (Q9UGN4-1) extracellular domain (Met 1-Gln 178) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8018