Learn More
Sino Biological Recombinant Human PHPT1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12473H07E100
Encompass_Preferred
A DNA sequence encoding the human PHPT1 (Q9NRX4-1) (Ala 2-Tyr 125) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8025