Learn More
Sino Biological Recombinant Human PLA2G2D Protein (hFc Tag)
Supplier:  Sino Biological 12479H02H100
Encompass_Preferred
A DNA sequence encoding the human PLA2G2D (Q9UNK4) (Met1-Cys145) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8031