Learn More
Sino Biological Recombinant Human CALML3 Protein (His Tag)
Supplier:  Sino Biological 12480H07E100
Encompass_Preferred
A DNA sequence encoding the human CALML3 (P27482) (Met 1-Lys 149) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8033