Learn More
Sino Biological Recombinant Human Ferritin light chain Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12482H07E100
Encompass_Preferred
A DNA sequence encoding the human FTL (P02792) (Met 1-Asp 175) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8035