Learn More
Sino Biological Recombinant Human BNIP3L Protein
Supplier:  Sino Biological 12488HNCE100
Encompass_Preferred
A DNA sequence encoding the human BNIP3L (NP_004322.1) (Ser 2-Lys 187) was expressed and purified, with additional two amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-8039