Learn More
Research Products International Corp 125/150 ml Flask Clamp for use with Mini-300 Orbital-Genie
SDP
Catalog No. 501978380
Encompass
Flask Clamps are for use with the Mini-300 Orbital-Genie and are made of stainless steel for added strength and safety. Mix and match different size clamps on same platform or use all same size. Includes mounting screws.
Note: Flask Clamps can also be used on the Orbital-Genie model with Adhering Mat by removing the blue Adhering Mat.
Catalog No. 50-197-8380 Supplier Research Products International Corp Supplier No. 400839
Flask Clamps are for use with the Mini-300 Orbital-Genie and are made of stainless steel for added strength and safety. Mix and match different size clamps on same platform or use all same size. Includes mounting screws.
Note: Flask Clamps can also be used on the Orbital-Genie model with Adhering Mat by removing the blue Adhering Mat.