Learn More
Waters Corp Waters Corporation Phosphate as PO4 Standard, 1000 mg/L, 125 mL
Catalog No. 50465869 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • This 1000 mg/L standard is for primary calibration or preparing second source calibration check standards.
  • It is traceable to NIST Standard Reference Materials, where available.
  • Guaranteed stable for one year.
  • Specifically designed for use on LC systems.
  • Helps establish calibration curves.
  • Increases analyst confidence.
  • Improves workflow efficiency.
  • Can assist in identifying matrix interferences.
  • Can be used to identify phosphate or phosphoric acid levels in soft beverages.
Catalog No. 50-465-869 Supplier Waters Corp Supplier No. 060
  • This 1000 mg/L standard is for primary calibration or preparing second source calibration check standards.
  • It is traceable to NIST Standard Reference Materials, where available.
  • Guaranteed stable for one year.
  • Specifically designed for use on LC systems.
  • Helps establish calibration curves.
  • Increases analyst confidence.
  • Improves workflow efficiency.
  • Can assist in identifying matrix interferences.
  • Can be used to identify phosphate or phosphoric acid levels in soft beverages.