Learn More
Chemglass Life Sciences 125mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 2mm Stpk
SDP
Catalog No. CG1742G03
Encompass_Preferred
125mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 2mm Stpk. Similar to CG-1742, but supplied without the PTFE stopcock plug or the flask-length stopper.
Catalog No. CG1742G03 Supplier Chemglass Life Sciences Supplier No. CG1742G03
Add to Cart
Add to Cart
125mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 2mm Stpk. Similar to CG-1742, but supplied without the PTFE stopcock plug or the flask-length stopper.