Learn More
Integra Biosciences Corp Non Sterile, Unfiltered, Low Retention, 125 ul Standard Tips, 5 XYZ Racks of 384 tips, 1920/CS
SDP
Catalog No. NC1399048
Encompass
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are  at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.
Catalog No. NC1399048 Supplier Integra Biosciences Corp Supplier No. 6563
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are  at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.