Learn More
Sino Biological Recombinant Human BVES Protein (GST Tag)
Supplier:  Sino Biological 12508H09E100
Encompass_Preferred
A DNA sequence encoding the human BVES (NP_671488.1) (Met1-Asn36) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8043