Learn More
Pacer Global LLC 1000 μL Extended Length Universal Pipette Tips, Filtered, Sterile, Low Retention, 96 tips per rack, 10 racks per pack
SDP
Supplier:  Pacer Global LLC MLSPTFS1000/DEL
Encompass_Preferred
1000 μL Extended Length Universal Pipette Tips, Filtered, Sterile, Low Retention, 96 tips per rack, 10 racks per pack. Lot traceable and DNase free, RNase free, and non-pyrogenic. Additional info on MLS-PTFS-1000: Sterile & Super low retention. Made of clear, high-quality polypropylene. Molded volume graduation markings. Interior support ribs prevent rack from warping. Smooth, hydrophobic surfaces to ensure accuracy. Lids have a stacking feature for stability and a secure locking tab. Packaged in blue rigid autoclavable racks. Filters reduce cross contamination and prevent damaging aerosols from penetrating into the pipette. MADE IN THE USA. Certifications: ISO13485:2016& ISO9001:2015. These pipette tips are universal fit and are compatible with major pipettes, including Eppendorf and Corning.
Catalog No. NC2609875