Learn More
Integra Biosciences Corp 1250 μl GripTips Sterile Filter, 10 Eco-Racks of 96 Tips, Low Retention
SDP
Catalog No. NC2387055
Encompass
The Integra 1250 μL Griptips Sterile Filter Tips are low retention pipette tips designed for precise liquid handling. They come in 10 eco-racks of 96 tips each, for a total of 960 tips. The tips have a maximum volume of 1250 μL and feature a hydrophobic PE filter membrane for protection against aerosols. Made of medical grade virgin polypropylene, these sterile tips have low binding properties to reduce sample loss. With their wide opening and smooth inner walls, they allow easy aspiration and dispensing. The slim design provides good visibility and access. These filter tips are ideal for PCR set-up and other sensitive applications requiring sterile conditions.
Catalog No. NC2387055 Supplier Integra Biosciences Corp Supplier No. 3845
The Integra 1250 μL Griptips Sterile Filter Tips are low retention pipette tips designed for precise liquid handling. They come in 10 eco-racks of 96 tips each, for a total of 960 tips. The tips have a maximum volume of 1250 μL and feature a hydrophobic PE filter membrane for protection against aerosols. Made of medical grade virgin polypropylene, these sterile tips have low binding properties to reduce sample loss. With their wide opening and smooth inner walls, they allow easy aspiration and dispensing. The slim design provides good visibility and access. These filter tips are ideal for PCR set-up and other sensitive applications requiring sterile conditions.