Learn More
Bellco Glass, Inc. 1250µL Low Retention Pipette Tips, Ovation, VistaRak, Non-sterile
SDP
Catalog No. 502329879
Encompass_Preferred
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time. VistaRak - Low RetentionLow Retention pipette tips are an ideal choice when working with viscous and low surface tension liquids to improve pipetting performance. Low retention tips work well with detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween.•Improved sample accuracy•Saves costly reagents or enzymes•Reduced sample loss for higher reproducibility•Unaffected by detergents used in sample buffers and reagents•Available sterile and non-sterile•Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved•Rack features 16 x 12 multichannel layout – 192 tips in a hinged box with a footprint of just 4.5” x 6”
Catalog No. 50-232-9879 Supplier Bellco Glass, Inc. Supplier No. 451900171
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time. VistaRak - Low RetentionLow Retention pipette tips are an ideal choice when working with viscous and low surface tension liquids to improve pipetting performance. Low retention tips work well with detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween.•Improved sample accuracy•Saves costly reagents or enzymes•Reduced sample loss for higher reproducibility•Unaffected by detergents used in sample buffers and reagents•Available sterile and non-sterile•Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved•Rack features 16 x 12 multichannel layout – 192 tips in a hinged box with a footprint of just 4.5” x 6”