Learn More
Santa Cruz Biotechnology EZ Touch Standard Pipette Tips in Sterile Racks, 50-1250 uL, 8 racks/case
Catalog No. NC1839626
Encompass
EZ Touch Pipette Tips are low binding, easy to seal and easy to eject, making them ideal for multi-channel pipettors. EZ Touch tips feature a soft, leak-proof top that allows for less pressure to create an optimal seal and minimal pressure to eject tips, thus reducing wear on pipettors.
Catalog No. NC1839626 Supplier Santa Cruz Biotechnology Supplier No. SC357415
EZ Touch Pipette Tips are low binding, easy to seal and easy to eject, making them ideal for multi-channel pipettors. EZ Touch tips feature a soft, leak-proof top that allows for less pressure to create an optimal seal and minimal pressure to eject tips, thus reducing wear on pipettors.