Learn More
Bellco Glass, Inc. 1250ul Pipette Tips, Clear, Non-Sterile, Bulk, Non-Sterile, 1000/pk, 10000/cs
SDP
Catalog No. 502329539
Encompass_Preferred
1250ul Pipette Tips, Clear, Non-Sterile, Bulk, Low-retention, Non-Sterile, 1000/pk, 10000/cs. Pipette Tips are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipette Tip line is packaged Rnase/Dnase, pyrogen free. Choose the right size for your research: 10µl, 100µl, 200µl, 300µl or 1000µl volumes. • Certified RNase, DNase, Human DNA, Pyrogen-Free.• Low-Retention, Fine-Point design for lowest sample adhesion.• Ultra-Clear polymer withstands organic solvents and provides clear, error-free pipetting.• Effective hydrophobic aerosol barrier eliminates carry-over and pipettor contamination.• Gamma-irradiated Sterile and Graduated.• Universal-fit for most single and multichannel pipettors.
Catalog No. 50-232-9539 Supplier Bellco Glass, Inc. Supplier No. 451400312
1250ul Pipette Tips, Clear, Non-Sterile, Bulk, Low-retention, Non-Sterile, 1000/pk, 10000/cs. Pipette Tips are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipette Tip line is packaged Rnase/Dnase, pyrogen free. Choose the right size for your research: 10µl, 100µl, 200µl, 300µl or 1000µl volumes. • Certified RNase, DNase, Human DNA, Pyrogen-Free.• Low-Retention, Fine-Point design for lowest sample adhesion.• Ultra-Clear polymer withstands organic solvents and provides clear, error-free pipetting.• Effective hydrophobic aerosol barrier eliminates carry-over and pipettor contamination.• Gamma-irradiated Sterile and Graduated.• Universal-fit for most single and multichannel pipettors.