Learn More
Bellco Glass, Inc. 1250µL Pipette Tips, Ovation, Filtered, VistaRak, RNase/DNase Free & Non-Pyrogenic Certified, Sterile
SDP
Supplier:  Bellco Glass, Inc. 451900193
Encompass_Preferred
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time. VistaRak - FilteredFilters reduce cross contamination and prevent aerosols from penetrating into the pipetteSterile onlyTips and racks are manufactured from 100% virgin polypropylene and can be autoclavedRack features 16 x 12 multichannel layout – 192 tips in a hinged box with a footprint of just 4.5” x 6”
Catalog No. 50-233-0834