Learn More
Sino Biological Recombinant Human ALDH3A1 Protein (His Tag)
Supplier:  Sino Biological 12523H07B50
Encompass_Preferred
A DNA sequence encoding the human ALDH3A1 (AAH04370.1) (Met 1-His 453) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8048