Learn More
Sino Biological Recombinant Human GLT25D2 Protein (His Tag)
Supplier:  Sino Biological 12542H08B100
Encompass_Preferred
A DNA sequence encoding the human GLT25D2 (Q8IYK4) (Met 1-Ser 622) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8049