Learn More
Sino Biological Recombinant Human p19 INK4d Protein (GST Tag)
Supplier:  Sino Biological 12558H09E50
Encompass_Preferred
A DNA sequence encoding the human CDKN2D (P55273) (Met 10Leu 166) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8058