Learn More
Sino Biological Recombinant Human GBP1 Protein (His Tag)
Supplier:  Sino Biological 12569H08H100
Encompass_Preferred
A DNA sequence encoding the human GBP1 (AAA35871.1) (Met 1-Cys 589) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8059