Learn More
Sino Biological Recombinant Human GAD67 Protein (His Tag)
Supplier:  Sino Biological 12570H08B50
Encompass_Preferred
A DNA sequence encoding the human GAD1 (Q99259-1) (Met 1-Leu 594) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8062